siRNA Details
| Gene Symbol: Htr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024395.1 | NP_077371.1 | 2 | 1350 | ||
| GenBank mRNA | L10073.1 | - | 2 | 1353 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr5b| Gene Symbol: Htr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024395.1 | NP_077371.1 | 2 | 1350 | ||
| GenBank mRNA | L10073.1 | - | 2 | 1353 | ||
Set Membership: |
|