siRNA Details
Gene Symbol: Htr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024395.1 | NP_077371.1 | 1 | 554 | ||
GenBank mRNA | L10073.1 | - | 1 | 557 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
