siRNA Details
| Gene Symbol: Abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024397.2 | NP_077373.2 | 5 | 567 | ||
| XM_006254353.2 | XP_006254415.1 | 5 | 638 | |||
| XM_006254354.2 | XP_006254416.1 | 4 | 622 | |||
| XM_006254355.2 | XP_006254417.1 | 4 | 622 | |||
| XM_006254356.2 | XP_006254418.1 | 5 | 637 | |||
| XM_006254357.2 | XP_006254419.1 | 4 | 622 | |||
| XM_006254358.2 | XP_006254420.1 | 5 | 637 | |||
| XM_006254359.2 | XP_006254421.1 | 4 | 622 | |||
| XM_006254360.2 | XP_006254422.1 | 5 | 655 | |||
| GenBank mRNA | AF176784.1 | - | 4 | 489 | ||
| BC129092.1 | - | 5 | 523 | 7192971 | ||

Abi1
Genomic Map