siRNA Details
| Gene Symbol: Aspa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024399.1 | NP_077375.1 | 1 | 276 | ||
| XM_006246815.2 | XP_006246877.1 | 1 | 318 | |||
| XM_006246817.2 | XP_006246879.1 | 1 | 318 | |||
| XM_006246818.2 | XP_006246880.1 | 1 | 318 | |||
| GenBank mRNA | AB023432.1 | - | 1 | 276 | ||
| BC078813.1 | - | 1 | 278 | 7129010 | ||
| BC078814.1 | - | 498 | 7133944 | |||
| FQ232103.1 | - | 1 | 276 | |||

Aspa
Genomic Map