siRNA Details
| Gene Symbol: Axin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024405.1 | NP_077381.1 | 1982 | |||
| GenBank mRNA | AF017756.1 | - | 1982 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Axin1| Gene Symbol: Axin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024405.1 | NP_077381.1 | 1982 | |||
| GenBank mRNA | AF017756.1 | - | 1982 | |||