siRNA Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053327.1 | NP_445779.1 | 16 | 1678 | ||
| XM_008764291.1 | XP_008762513.1 | 16 | 1794 | |||
| XM_008764292.1 | XP_008762514.1 | 16 | 1764 | |||
| GenBank mRNA | BC081761.1 | - | 16 | 1744 | 7108273 | |
| D13927.1 | - | 16 | 1678 | |||
| Z34291.1 | - | 15 | 1635 | |||

Clcnka
Genomic Map