siRNA Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053327.1 | NP_445779.1 | 10 | 973 | ||
| GenBank mRNA | D13927.1 | - | 10 | 973 | ||
| Z34291.1 | - | 9 | 930 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcnka| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053327.1 | NP_445779.1 | 10 | 973 | ||
| GenBank mRNA | D13927.1 | - | 10 | 973 | ||
| Z34291.1 | - | 9 | 930 | |||