siRNA Details
Gene Symbol: Luzp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030830.2 | NP_110457.2 | 4 | 2629 | ||
XM_006239274.2 | XP_006239336.1 | 3 | 2572 | |||
GenBank mRNA | AF181259.1 | - | 5 | 4579 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Luzp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030830.2 | NP_110457.2 | 4 | 2629 | ||
XM_006239274.2 | XP_006239336.1 | 3 | 2572 | |||
GenBank mRNA | AF181259.1 | - | 5 | 4579 |