siRNA Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031317.1 | NP_112607.1 | 3 | 344 | ||
| GenBank mRNA | AB033830.1 | - | 3 | 344 | ||
| AF508348.1 | - | 2 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfc| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031317.1 | NP_112607.1 | 3 | 344 | ||
| GenBank mRNA | AB033830.1 | - | 3 | 344 | ||
| AF508348.1 | - | 2 | 222 | |||