siRNA Details
| Gene Symbol: Timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032618.1 | NP_116007.1 | 1 | 177 | ||
| GenBank mRNA | AF223951.1 | - | 1 | 161 | ||
| FM107575.1 | - | 1 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm22| Gene Symbol: Timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032618.1 | NP_116007.1 | 1 | 177 | ||
| GenBank mRNA | AF223951.1 | - | 1 | 161 | ||
| FM107575.1 | - | 1 | 177 | |||