siRNA Details
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024486.1 | NP_077812.1 | 3, 4 | 474 | ||
| XM_006234214.2 | XP_006234276.1 | 5, 6 | 2079 | |||
| XM_008761862.1 | XP_008760084.1 | 6, 7 | 2136 | |||
| XM_008761863.1 | XP_008760085.1 | 6, 7 | 4380 | |||
| XM_008761864.1 | XP_008760086.1 | 6, 7 | 4126 | |||
| XM_008761865.1 | XP_008760087.1 | 6, 7 | 4169 | |||
| XM_008761866.1 | XP_008760088.1 | 6, 7 | 846 | |||
| XM_008761867.1 | XP_008760089.1 | 3, 4 | 390 | |||
| GenBank mRNA | BC167754.1 | - | 4, 5 | 645 | 9089038 | |
| L19341.1 | - | 3, 4 | 474 | |||

Acvr1
Genomic Map