siRNA Details
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024486.1 | NP_077812.1 | 7 | 1197 | ||
| XM_006234214.2 | XP_006234276.1 | 9 | 2802 | |||
| XM_008761862.1 | XP_008760084.1 | 10 | 2859 | |||
| XM_008761863.1 | XP_008760085.1 | 10 | 5103 | |||
| XM_008761864.1 | XP_008760086.1 | 10 | 4849 | |||
| XM_008761865.1 | XP_008760087.1 | 10 | 4892 | |||
| XM_008761866.1 | XP_008760088.1 | 10 | 1569 | |||
| XM_008761867.1 | XP_008760089.1 | 7 | 1113 | |||
| GenBank mRNA | BC167754.1 | - | 8 | 1368 | 9089038 | |
| L19341.1 | - | 7 | 1197 | |||

Acvr1
Genomic Map