siRNA Details
| Gene Symbol: Zbtb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024489.1 | NP_077815.1 | 3 | 1833 | ||
| GenBank mRNA | AF091457.1 | - | 3 | 1833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb10| Gene Symbol: Zbtb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024489.1 | NP_077815.1 | 3 | 1833 | ||
| GenBank mRNA | AF091457.1 | - | 3 | 1833 | ||