siRNA Details
Gene Symbol: Inpp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031002.3 | NP_112264.3 | 20 | 2233 | ||
GenBank mRNA | CB614071.1 | - | 4 | 413 | ||
U26397.1 | - | 20 | 2233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Inpp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031002.3 | NP_112264.3 | 20 | 2233 | ||
GenBank mRNA | CB614071.1 | - | 4 | 413 | ||
U26397.1 | - | 20 | 2233 |