siRNA Details
| Gene Symbol: Slc16a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030834.1 | NP_110461.1 | 5 | 1245 | ||
| XM_006247931.2 | XP_006247993.1 | 6 | 1360 | |||
| XM_006247932.2 | XP_006247994.1 | 6 | 1652 | |||
| XM_006247933.2 | XP_006247995.1 | 5 | 1267 | |||
| XM_006247934.2 | XP_006247996.1 | 6 | 1326 | |||
| XM_008768513.1 | XP_008766735.1 | 6 | 4084 | |||
| XM_008768514.1 | XP_008766736.1 | 5 | 3991 | |||
| GenBank mRNA | BC168146.1 | - | 5 | 1204 | 9089640 | |
| U87627.1 | - | 5 | 1245 | |||

Slc16a3
Genomic Map