siRNA Details
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030834.1 | NP_110461.1 | 4 | 950 | ||
GenBank mRNA | U87627.1 | - | 4 | 950 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030834.1 | NP_110461.1 | 4 | 950 | ||
GenBank mRNA | U87627.1 | - | 4 | 950 |