siRNA Details
| Gene Symbol: Syt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030839.3 | NP_110466.2 | 6 | 1221 | ||
| XM_008762036.1 | XP_008760258.1 | 6 | 1815 | |||
| GenBank mRNA | AF313453.1 | - | 6 | 1406 | ||
| AF375466.1 | - | 6 | 1221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt13| Gene Symbol: Syt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030839.3 | NP_110466.2 | 6 | 1221 | ||
| XM_008762036.1 | XP_008760258.1 | 6 | 1815 | |||
| GenBank mRNA | AF313453.1 | - | 6 | 1406 | ||
| AF375466.1 | - | 6 | 1221 | |||