siRNA Details
| Gene Symbol: Bdkrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030851.1 | NP_110478.1 | 2 | 691 | ||
| XM_008764861.1 | XP_008763083.1 | 1 | 569 | |||
| GenBank mRNA | AJ132230.2 | - | 2 | 691 | ||
| U66107.2 | - | 2 | 700 | |||
More Information
Set Membership: |
|

Bdkrb1
Genomic Map