siRNA Details
| Gene Symbol: Lrrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030856.1 | NP_110483.1 | 2 | 1661 | ||
| GenBank mRNA | AF291437.1 | - | 2 | 1661 | ||
| BC081791.1 | - | 1 | 1007 | 7124169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrn3| Gene Symbol: Lrrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030856.1 | NP_110483.1 | 2 | 1661 | ||
| GenBank mRNA | AF291437.1 | - | 2 | 1661 | ||
| BC081791.1 | - | 1 | 1007 | 7124169 | ||