siRNA Details
| Gene Symbol: Lyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111098.1 | NP_001104568.1 | 7 | 694 | ||
| NM_030857.2 | NP_110484.1 | 7 | 757 | |||
| XM_006237834.2 | XP_006237896.1 | 7 | 1073 | |||
| GenBank mRNA | AF000300.1 | - | 7 | 602 | ||
| AF000301.1 | - | 7 | 694 | |||
| AF000302.1 | - | 7 | 692 | |||
| L14782.1 | - | 6 | 551 | |||
| L14823.1 | - | 6 | 488 | |||
| L14951.1 | - | 7 | 760 | |||

Lyn
Genomic Map