siRNA Details
| Gene Symbol: Mgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030861.1 | NP_110488.1 | 2 | 1072 | ||
| XM_006246182.2 | XP_006246244.1 | 3 | 1550 | |||
| XM_006246183.2 | XP_006246245.1 | 2 | 1724 | |||
| XM_006246184.2 | XP_006246246.1 | 3 | 1831 | |||
| XM_006246185.2 | XP_006246247.1 | 4 | 1657 | |||
| XM_008767715.1 | XP_008765937.1 | 3 | 1564 | |||
| GenBank mRNA | BC074010.1 | - | 2 | 1096 | 7100030 | |
| D16302.1 | - | 2 | 1072 | |||

Mgat1
Genomic Map