siRNA Details
| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030863.1 | NP_110490.1 | 6 | 767 | ||
| GenBank mRNA | AF004811.1 | - | 6 | 767 | ||
| FQ230914.1 | - | 854 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msn| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030863.1 | NP_110490.1 | 6 | 767 | ||
| GenBank mRNA | AF004811.1 | - | 6 | 767 | ||
| FQ230914.1 | - | 854 | ||||