siRNA Details
Gene Symbol: Nrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030869.3 | NP_110496.1 | 6 | 1137 | ||
XM_006245039.2 | XP_006245101.1 | 6 | 1695 | |||
XM_006245040.2 | XP_006245102.1 | 6 | 1694 | |||
XM_006245041.2 | XP_006245103.1 | 6 | 1696 | |||
XM_006245042.2 | XP_006245104.1 | 6 | 1695 | |||
XM_008767137.1 | XP_008765359.1 | 6 | 1694 | |||
GenBank mRNA | AF016297.1 | - | 6 | 1170 |
