siRNA Details
| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030871.1 | NP_110498.1 | 158 | |||
| GenBank mRNA | AF327836.1 | - | 158 | |||
| AF327838.1 | - | 158 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde1a| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030871.1 | NP_110498.1 | 158 | |||
| GenBank mRNA | AF327836.1 | - | 158 | |||
| AF327838.1 | - | 158 | ||||