siRNA Details
| Gene Symbol: Ceacam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033860.1 | NP_001029032.1 | 2 | 409 | ||
| NM_001033861.1 | NP_001029033.1 | 2 | 409 | |||
| NM_001033862.1 | NP_001029034.1 | 2 | 409 | |||
| NM_031755.2 | NP_113943.1 | 2 | 409 | |||
| GenBank mRNA | AJ277104.1 | - | 2 | 371 | ||
| AJ277105.1 | - | 2 | 371 | |||
| J04963.1 | - | 2 | 416 | |||
| M92848.1 | - | 2 | 409 | |||
| X91137.1 | - | 2 | 371 | |||

Ceacam1
Genomic Map