siRNA Details
Gene Symbol: Abat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031003.2 | NP_112265.1 | 13 | 1113 | ||
XM_006245760.2 | XP_006245822.1 | 13 | 1321 | |||
GenBank mRNA | BC081787.1 | - | 13 | 1110 | 7109775 | |
D87839.1 | - | 13 | 1114 | |||
U29701.1 | - | 12 | 1012 |
