siRNA Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031003.2 | NP_112265.1 | 13 | 1160 | ||
| XM_006245760.2 | XP_006245822.1 | 13 | 1368 | |||
| GenBank mRNA | BC081787.1 | - | 13 | 1157 | 7109775 | |
| D87839.1 | - | 13 | 1161 | |||
| U29701.1 | - | 12 | 1059 | |||

Abat
Genomic Map