siRNA Details
| Gene Symbol: Adar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031006.1 | NP_112268.1 | 15 | 3478 | ||
| XM_006232774.2 | XP_006232836.1 | 16 | 3702 | |||
| XM_006232775.2 | XP_006232837.1 | 16 | 3623 | |||
| XM_006232776.2 | XP_006232838.1 | 15 | 6586 | |||
| XM_006232777.2 | XP_006232839.1 | 15 | 3510 | |||
| XM_006232778.2 | XP_006232840.1 | 15 | 3462 | |||
| GenBank mRNA | FQ212262.1 | - | 3685 | |||
| FQ212270.1 | - | 3685 | ||||
| U18942.1 | - | 15 | 3478 | |||

Adar
Genomic Map