siRNA Details
| Gene Symbol: Ap2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031008.2 | NP_112270.2 | 9 | 1191 | ||
| XM_008760010.1 | XP_008758232.1 | 9 | 1246 | |||
| GenBank mRNA | BC081786.1 | - | 9 | 1191 | 7126468 | |
| X53773.1 | - | 1144 | ||||

Ap2a2
Genomic Map