siRNA Details
| Gene Symbol: Anpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031012.1 | NP_112274.1 | 14, 15 | 2176 | ||
| XM_006229407.2 | XP_006229469.1 | 15, 16 | 2596 | |||
| XM_006229408.2 | XP_006229470.1 | 15, 16 | 2617 | |||
| XM_006229409.1 | XP_006229471.1 | 15, 16 | 2653 | |||
| GenBank mRNA | M25073.1 | - | 14, 15 | 2176 | ||
| M26710.1 | - | 14, 15 | 2223 | |||
| Y18765.1 | - | 14, 15 | 1566 | |||

Anpep
Genomic Map