siRNA Details
| Gene Symbol: Atic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031014.2 | NP_112276.2 | 12 | 1249 | ||
| GenBank mRNA | BC072496.1 | - | 12 | 1249 | 7096880 | |
| D89514.1 | - | 12 | 1242 | |||
| Gene Symbol: LOC100910688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750715.3 | XP_003750763.1 | 319 | |||

Atic
Genomic Map