siRNA Details
| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031016.1 | NP_112278.1 | 1 | 614 | ||
| GenBank mRNA | AB017655.1 | - | 1 | 163 | ||
| J03025.1 | - | 1 | 614 | |||
More Information
Other Species Targeted
Set Membership: |
|

Chrm2
Genomic Map