siRNA Details
Gene Symbol: Chrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031016.1 | NP_112278.1 | 1 | 1090 | ||
GenBank mRNA | AB017655.1 | - | 1 | 639 | ||
J03025.1 | - | 1 | 1090 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
