siRNA Details
| Gene Symbol: Cspg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031022.1 | NP_112284.1 | 10 | 5254 | ||
| GenBank mRNA | X56541.2 | - | 10 | 5254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cspg4| Gene Symbol: Cspg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031022.1 | NP_112284.1 | 10 | 5254 | ||
| GenBank mRNA | X56541.2 | - | 10 | 5254 | ||