siRNA Details
| Gene Symbol: Ctbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031023.1 | NP_112285.1 | 3 | 310 | ||
| GenBank mRNA | BC088129.1 | - | 3 | 313 | 7323494 | |
| FQ219359.1 | - | 3 | 319 | |||
| M95768.1 | - | 3 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctbs| Gene Symbol: Ctbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031023.1 | NP_112285.1 | 3 | 310 | ||
| GenBank mRNA | BC088129.1 | - | 3 | 313 | 7323494 | |
| FQ219359.1 | - | 3 | 319 | |||
| M95768.1 | - | 3 | 310 | |||