siRNA Details
| Gene Symbol: Dync1li2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031026.1 | NP_112288.1 | 2 | 170 | ||
| XM_006255053.2 | XP_006255115.1 | 2 | 384 | |||
| XM_006255054.2 | XP_006255116.1 | 2 | 384 | |||
| GenBank mRNA | BC089794.1 | - | 2 | 191 | 7115029 | |
| U15138.1 | - | 2 | 170 | |||

Dync1li2
Genomic Map