siRNA Details
| Gene Symbol: Gabbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031028.3 | NP_112290.2 | 9 | 1247 | ||
| XM_006255879.2 | XP_006255941.1 | 9 | 1341 | |||
| XM_008772700.1 | XP_008770922.1 | 8 | 1320 | |||
| GenBank mRNA | AB016160.1 | - | 4 | 790 | ||
| AB016161.1 | - | 4 | 790 | |||
| AF283276.1 | - | 8 | 1161 | |||
| AJ831471.1 | - | 9 | 1245 | |||
| AJ831472.1 | - | 9 | 1369 | |||
| Y10369.2 | - | 9 | 1219 | |||
| Y10370.2 | - | 4 | 892 | |||
More Information
Set Membership: |
|

Gabbr1
Genomic Map