siRNA Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031029.1 | NP_112291.1 | 6 | 610 | ||
| XM_008767594.1 | XP_008765816.1 | 7 | 1196 | |||
| XM_008767595.1 | XP_008765817.1 | 7 | 1202 | |||
| GenBank mRNA | FQ222085.1 | - | 7 | 777 | ||
| U95368.1 | - | 6 | 610 | |||

Gabrp
Genomic Map