siRNA Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031029.1 | NP_112291.1 | 8 | 1012 | ||
| XM_008767594.1 | XP_008765816.1 | 9 | 1598 | |||
| XM_008767595.1 | XP_008765817.1 | 9 | 1604 | |||
| GenBank mRNA | FQ222085.1 | - | 9 | 1179 | ||
| U95368.1 | - | 8 | 1012 | |||

Gabrp
Genomic Map