siRNA Details
Gene Symbol: Gak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031030.2 | NP_112292.1 | 20 | 2462 | ||
XM_006250583.2 | XP_006250645.1 | 20 | 2477 | |||
GenBank mRNA | D38560.1 | - | 20 | 2463 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031030.2 | NP_112292.1 | 20 | 2462 | ||
XM_006250583.2 | XP_006250645.1 | 20 | 2477 | |||
GenBank mRNA | D38560.1 | - | 20 | 2463 |