siRNA Details
| Gene Symbol: Gak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031030.2 | NP_112292.1 | 5 | 538 | ||
| XM_006250583.2 | XP_006250645.1 | 5 | 553 | |||
| XM_006250584.2 | XP_006250646.1 | 5 | 553 | |||
| GenBank mRNA | D38560.1 | - | 5 | 539 | ||

Gak
Genomic Map