siRNA Details
| Gene Symbol: Gak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031030.2 | NP_112292.1 | 23 | 3202 | ||
| XM_006250583.2 | XP_006250645.1 | 23 | 3136 | |||
| XM_006250584.2 | XP_006250646.1 | 22 | 3070 | |||
| GenBank mRNA | D38560.1 | - | 23 | 3203 | ||

Gak
Genomic Map