siRNA Details
| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031032.2 | NP_112294.1 | 7 | 464 | ||
| XM_006251814.2 | XP_006251876.1 | 7 | 1128 | |||
| GenBank mRNA | BC081778.1 | - | 7 | 464 | 7192655 | |
| FQ222145.1 | - | 7 | 526 | |||

Gmfb
Genomic Map