siRNA Details
Gene Symbol: Gnaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031036.1 | NP_112298.1 | 7 | 936 | ||
GenBank mRNA | AF234260.1 | - | 7 | 936 | ||
DQ120477.1 | - | 7 | 901 | |||
DQ120478.1 | - | 7 | 901 | |||
L37294.1 | - | 2 | 207 |
