siRNA Details
Gene Symbol: Gnrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031038.3 | NP_112300.2 | 3 | 960 | ||
GenBank mRNA | L07646.1 | - | 3 | 946 | ||
X68980.1 | - | 3 | 910 | |||
X76635.1 | - | 3 | 899 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
