siRNA Details
Gene Symbol: Grm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031040.1 | NP_112302.1 | 6 | 1738 | ||
XM_008763199.1 | XP_008761421.1 | 6 | 1738 | |||
GenBank mRNA | D16817.1 | - | 6 | 1738 | ||
U06832.1 | - | 6 | 1473 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
