siRNA Details
| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031041.1 | NP_112303.1 | 7 | 850 | ||
| GenBank mRNA | BC085345.1 | - | 7 | 855 | 7303747 | |
| FQ229769.1 | - | 7 | 839 | |||
| X65948.1 | - | 7 | 850 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf2b| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031041.1 | NP_112303.1 | 7 | 850 | ||
| GenBank mRNA | BC085345.1 | - | 7 | 855 | 7303747 | |
| FQ229769.1 | - | 7 | 839 | |||
| X65948.1 | - | 7 | 850 | |||