siRNA Details
| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031041.1 | NP_112303.1 | 5 | 483 | ||
| GenBank mRNA | BC085345.1 | - | 5 | 488 | 7303747 | |
| FQ229769.1 | - | 5 | 471 | |||
| FQ231337.1 | - | 5 | 513 | |||
| X65948.1 | - | 5 | 483 | |||

Gtf2b
Genomic Map