siRNA Details
| Gene Symbol: Itpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031046.3 | NP_112308.1 | 38 | 5516 | ||
| GenBank mRNA | AF329470.2 | - | 38 | 5491 | ||
| AY313846.1 | - | 38 | 5270 | |||
| X61677.1 | - | 38 | 5516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpr2| Gene Symbol: Itpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031046.3 | NP_112308.1 | 38 | 5516 | ||
| GenBank mRNA | AF329470.2 | - | 38 | 5491 | ||
| AY313846.1 | - | 38 | 5270 | |||
| X61677.1 | - | 38 | 5516 | |||