siRNA Details
| Gene Symbol: Mmp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031056.1 | NP_112318.1 | 3 | 446 | ||
| GenBank mRNA | BC072509.1 | - | 3 | 477 | 7103279 | |
| X83537.1 | - | 3 | 455 | |||
| X91785.1 | - | 3 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp14| Gene Symbol: Mmp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031056.1 | NP_112318.1 | 3 | 446 | ||
| GenBank mRNA | BC072509.1 | - | 3 | 477 | 7103279 | |
| X83537.1 | - | 3 | 455 | |||
| X91785.1 | - | 3 | 446 | |||